missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEA15 Monoclonal Antibody (4D2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547430
This item is not returnable.
View return policy
Description
PEA15 Monoclonal Antibody for Western Blot, ICC/IF, IHC (PFA)
PEA15 is a death effector domain (DED)-containing protein predominantly expressed in the central nervous system, particularly in astrocytes.
Specifications
| PEA15 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 5 mM sodium azide | |
| Q15121, Q5U318, Q62048 | |
| PEA15, Pea15a | |
| Full length human PEA-15 as expressed in and purified from E. coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
| 4D2 | |
| Unconjugated | |
| PEA15 | |
| 15 kDa phosphoprotein enriched in astrocytes; astrocytic phosphoprotein PEA-15; HMAT1; homolog of mouse MAT-1 oncogene; HUMMAT1H; mammary transforming gene 1; mammary transforming gene 1, mouse, homolog of; MAT1; MAT1H; OTTHUMP00000029887; Pea15; PEA-15; Pea15a; PED; phosphoprotein enriched in astrocytes 15; phosphoprotein enriched in astrocytes 15A; phosphoprotein enriched in diabetes; Pkcs15; proliferation and apoptosis adaptor protein 15 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 18611, 364052, 510586, 8682 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction