Learn More
PEAR1 Mouse anti-Human, Brilliant Violet 421, Clone: 492621, BD OptiBuild™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 748061
Product Code. 17209704
  • ONLINE EXCLUSIVE PRICE
    € 540.00
    € 500.00 / 50µg
    Save € 40.00
    7% Off
Description
PEAR1 Monoclonal antibody specifically detects PEAR1 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PEAR1
Monoclonal
0.2 mg/mL
Mouse
RUO
PEAR1
Refrigerate (2 to 8°C)
Flow Cytometry
492621
Brilliant Violet 421
50 μg
Primary
Human
IgG2a κ