Learn More
Invitrogen™ PEBP1 Recombinant Rabbit Monoclonal Antibody (24GB860)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557538
 View more versions of this product
Product Code. 30274851
  • € 580.00 / 100µL
Description
PEBP1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
PBP binds ATP, opioids and phosphatidylethanolamine, exhibiting a lower affinity for phosphatidylinositol and phosphatidylcholine. This serine protease inhibitor inhibits thrombin, neuropsin and chymotrypsin but not trypsin, tissue type plasminogen activator and elastase. PBP contains hippocampal cholinergic neurostimulating peptide (HCNP), which may be involved in the function of the presynaptic cholinergic neurons of the central nervous system. HCNP increases the production of choline acetyltransferase but not acetylcholinesterase.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PEBP1
Recombinant Monoclonal
Unconjugated
PEBP1
23 kDa morphine-binding protein; Basic cytosolic 21 kDa protein; epididymis luminal protein 210; epididymis secretory protein Li 34; epididymis secretory protein Li 96; HCNP; HCNPpp; HEL-210; HEL-S-34; HEL-S-96; Hippocampal cholinergic neurostimulating peptide; KIAA0299; LDGF; MDGF; MOCA; neuropolypeptide h3; P23K; PBP; Pbp1; Pbpr; PEBP; PEBP1; PEBP-1; phosphatidylethanolamine binding protein; phosphatidylethanolamine binding protein 1; phosphatidylethanolamine-binding protein; phosphatidylethanolamine-binding protein 1; PPBP; prostatic binding protein; Prostatic-binding protein; Raf kinase inhibitor protein; Raf kinase inhibitory protein; Raf-1 inhibitor protein; Raf-1 kinase inhibitor protein; RKIP; similar to phosphatidylethanolamine binding protein; zgc:56033; zgc:76942
Rabbit
Affinity Chromatography
RUO
23980, 29542, 5037
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB860
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P30086, P31044, P70296
PEBP1
A synthesized peptide derived from human PEBP1 (aa 64-99).