missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEBP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120598
This item is not returnable.
View return policy
Description
Positive test controls include: Human plasma, Rat testis. Immunogen sequence: RQKITAWQAP IVKFHTALDG ALYLLVMVDP DAPSRSNPVM KYWRHWLVSN ITGADMKSGS IRGNVLSDYS PPTPPPETGL HRYQFFVYLQ GDRDISLSVE EKADLGGWNL DKFLQQYGLR DPDTSTQFMT QFDEELSSEF GRINDDQEQF NQK.
The phosphatidylethanolamine -binding proteins, including PEBP4, are an evolutionarily conserved family of proteins with pivotal biologic functions, such as lipid binding and inhibition of serine proteases.
Specifications
| PEBP4 | |
| Polyclonal | |
| Unconjugated | |
| Pebp4 | |
| CORK1; CORK-1; cousin-of-RKIP 1 protein; epididymis secretory protein Li 300; GWTM1933; HEL-S-300; hPEBP4; MGC22776; PEBP4; PEBP-4; phosphatidylethanolamine binding protein 4; phosphatidylethanolamine-binding protein 4; PRO4408; Protein cousin-of-RKIP 1; UNQ1933/PRO4408 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 157310, 691997, 73523 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.66 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q96S96, Q9D9G2 | |
| Pebp4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 90-242 of mouse PEBP4 (NP_0828362). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction