missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCD1 Mouse anti-Human, Clone: OTI3A1, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803561S
This item is not returnable.
View return policy
Description
ABCD1 Monoclonal antibody specifically detects ABCD1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| ABCD1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P33897 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 215 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3A1 | |
| Unconjugated | |
| ABCD1 | |
| ABC42; ABCD1; Adrenoleukodystrophy protein; ALD; Aldgh; ALDP; AMN; ATP binding cassette subfamily D member 1; ATP-binding cassette sub-family D member 1; ATP-binding cassette, subfamily D (ALD), member 1; ATP-binding cassette, sub-family D (ALD), member 1; ATP-binding cassette, sub-family D, member 1; RGD1562128; X-linked adrenoleukodystrophy (ALD) gene homolog | |
| Human recombinant protein fragment corresponding to amino acids 508-745 of human ABCD1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction