missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PECAM1, Mouse anti-Human, Clone: 1H6, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB16410.50uL
This item is not returnable.
View return policy
Specifications
| PECAM1 | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.3 (50% glycerol, 1% BSA, 0.02% sodium azide). | |
| CD31/PECAM-1 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1H6 | |
| Flow Cytometry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| platelet/endothelial cell adhesion molecule | |
| PECAM1 | |
| Recombinant protein corresponding to full length human PECAM1 from HEK293T cell. | |
| RUO | |
| 5175 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction