missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PECI Monoclonal Antibody (K16256_4D6), Invitrogen™

Product Code. p-200088926
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
200µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30194735 200 μL 200µg
30211632 50 μL 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30194735 Supplier Invitrogen™ Supplier No. MA556324

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sequence of this protein is as follows: KDFENSMNQV KLLKKDPGNE VKLKLYALYK QATEGPCNMP KPGVFDLINK AKWDAWNALG SLPKEAARQN YVDLVSSLSP SLESSSQVEP GTDRKSTGFE TLVVTSEDGI TKIMFNRPKK KNAINTEMYH EIMRALKAAS KDDSIITVLT GNGDYYSSGN DLTNFTDIPP GGVEEKAKNN AVLLREFVGC FIDFPKPLIA VVNGPAVGIS VTLLGLFDAV YASDRATFHT PFSHLGQSPE GCSSYTFPKI MSPAKATEML IFGKKLTAGE ACAQGLVTEV FPDSTFQKEV WTRLKAFAKL PPNALRISKE VIRKREREKL HAVNAEECNV LQGRWLSDEC TNAVVNFLSR

PECI is an auxiliary enzyme that catalyzes an isomerization step required for the beta-oxidation of unsaturated fatty acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PECI
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K16256_4D6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene ECI2
Gene Accession No. O75521
Gene Alias ACBD2; acyl-Coenzyme A binding domain containing 2; D3,D2-enoyl-CoA isomerase; DBI-related protein 1; delta(3),delta(2)-enoyl-CoA isomerase; Diazepam-binding inhibitor-related protein 1; dJ1013A10.3; Dodecenoyl-CoA isomerase; Drs1; DRS-1; Eci2; enoyl-CoA delta isomerase 2; enoyl-CoA delta isomerase 2, mitochondrial; enoyl-Coenzyme A delta isomerase 2; Hca88; Hepatocellular carcinoma-associated antigen 88; PECI; Peroxisomal 3,2-trans-enoyl-CoA isomerase; peroxisomal 3,2-trans-enoyl-CoA isomerase; enoyl-CoA delta isomerase 2, mitochondrial; peroxisomal D3,D2-enoyl-CoA isomerase; peroxisomal delta3, delta2-enoyl-Coenzyme A isomerase; renal carcinoma antigen NY-REN-1; testicular secretory protein Li 33
Gene Symbols ECI2
Host Species Mouse
Immunogen Recombinant human ECI2 (immunogen range is 41-390).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10455
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.