missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PECR Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | PECR |
|---|---|
| Clone | 2D3G1 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30233479
|
Proteintech
68248-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234962
|
Proteintech
68248-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PECR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PECR. This antibody recognizes human, mouse, rat antigen. The PECR antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| PECR | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 55825 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 2D3G1 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| HSA250303; SDR29C1; TERP | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title