missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEDFR/PNPLA2/ATGL Antibody (AT18E6), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59390PE
This item is not returnable.
View return policy
Description
PEDFR/PNPLA2/ATGL Monoclonal antibody specifically detects PEDFR/PNPLA2/ATGL in Human samples. It is validated for Flow Cytometry
Specifications
| PEDFR/PNPLA2/ATGL | |
| Monoclonal | |
| PE | |
| Adipose triglyceride lipase, ATGL1110001C14Rik, Calcium-independent phospholipase A2, Desnutrin, EC 3.1.1.3, FP17548, IPLA2-zeta, patatin-like phospholipase domain containing 2, patatin-like phospholipase domain-containing protein 2, patatin-like phospholipase domain-containing protein 2-like, PEDF-R, Pigment epithelium-derived factor, Transport-secretion protein 2, transport-secretion protein 2.2, triglyceride hydrolase, TTS2.2, TTS-2.2, TTS2DKFZp667M109 | |
| Recombinant human PEDFR/PNPLA2/ATGL (30-504aa) purified from E. coli | |
| 0.1 mL | |
| Autophagy, Cancer, Diabetes Research, Lipid and Metabolism, Lipid Droplets, Signal Transduction | |
| 57104 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Flow Cytometry | |
| AT18E6 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction