missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PEDFR/PNPLA2/ATGL Polyclonal specifically detects PEDFR/PNPLA2/ATGL in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | PEDFR/PNPLA2/ATGL |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/Immunofluorescence 1-4 ug/ml |
| Formulation | PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide |
| Gene Alias | Adipose triglyceride lipase, ATGL1110001C14Rik, Calcium-independent phospholipase A2, Desnutrin, EC 3.1.1.3, FP17548, IPLA2-zeta, patatin-like phospholipase domain containing 2, patatin-like phospholipase domain-containing protein 2, patatin-like phospholipase domain-containing protein 2-like, PEDF-R, Pigment epithelium-derived factor, Transport-secretion protein 2, transport-secretion protein 2.2, triglyceride hydrolase, TTS2.2, TTS-2.2, TTS2DKFZp667M109 |
| Gene Symbols | PNPLA2 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FSGESDICPQDSSTNIHELRVTNTSIQFNLRNLYRLSKALFPPEPLVLREMCKQGYRDGLRFLQRNGLLNRPNPLLALPPARPHGPEDKDQA |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?