Learn More
Hamilton™ PEEK Replacement Guard Cartridge
Product Code. 10698135
Change view
Click to view available options
Diameter (Metric) Inner:
3 mm
missing translation for 'unitSize'
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
10698135 3 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Product Code. 10698135 missing translation for 'mfr' Hamilton™ missing translation for 'supplierNo' 79385
Only null left
Add to basket
Add to basket
A more rugged column with exceptionally higher separation efficiencies than earlier predecessors.
  • Well suited for use in systems employing suppressed/non-suppressed conductivity, electrochemical, UV, and ICP-MS detection
  • Helps in reducing sample pretreatment
  • Helps improve the accuracy and precision of results
Specifications
Type PEEK Replacement Guard Cartridge
Diameter (Metric) Inner 3 mm