Learn More
Trajan Scientific and Medical PEEK Tubing, 1/8 in. O.D., 50 ft
Click to view available options
Diameter (English) Inner:
0.062 in.
0.080 in.
Unit Size:
Each
Description
- Excellent chemical resistance and inert to most commonly used solvents. Exceptions are very caustic reagents, strong acids and bases
- Bio-inert ad bio-compatible
- Color coded to industry standards for easy identification of inner diameter
- Leach-free colorants
- Smooth inner walls and precise IDs
- Works well with either polymer fittings as well as stainless steel fittings
- Can be formed to specific customer needs
Specifications
Specifications
| Color | Natural |
| pH Range | 2 to 14 |
| Diameter (English) Inner | 0.080 in. |
| Diameter (English) Outer | 1/8 in. |
| Overall Length (English) | 50 ft. |
| Material | PEEK |
| Maximum Pressure | 3000 psi (207 bar) |
| Max. Operating Temperature (Metric) | 100°C |