Learn More
PEF1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10151-1-AP-20UL
This item is not returnable.
View return policy
Description
PEF1 is a Ca(2+)-binding protein that belongs to the penta-EF-hand (PEF) protein family, which includes the calpain small subunit (CAPNS1), sorcin (SRI), grancalcin (GCA), and ALG2. PEF1 is a Ca(2+)-binding protein that belongs to the penta-EF-hand (PEF) protein family, which includes the calpain small subunit (CAPNS1), sorcin (SRI), grancalcin (GCA), and ALG2 (PDCD6) (Kitaura et al., 2001 ).Specifications
| PEF1 | |
| Polyclonal | |
| Unconjugated | |
| PEF1 | |
| ABP32, PEF1, PEF1A, PEFLIN | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 553115 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9UBV8 | |
| PEF1 | |
| PEF1 Fusion Protein Ag0198 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |