missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peflin Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00553115-B01P
This item is not returnable.
View return policy
Description
Peflin Polyclonal antibody specifically detects Peflin in Human samples. It is validated for Western Blot
Specifications
| Peflin | |
| Polyclonal | |
| PBS (pH 7.4) | |
| ABP32, PEF protein with a long N-terminal hydrophobic domain, PEF1A, peflin, penta-EF hand domain containing 1, Penta-EF hand domain-containing protein 1, penta-EF-hand domain containing 1 | |
| PEF1 (AAH02773, 1 a.a. - 284 a.a.) full-length human protein. MASYPYRQGCPGAAGQAPGAPPGSYYPGPPNSGGQYGSGLPPGGGYGGPAPGGPYGPPAGGGPYGHPNPGMFPSGTPGGPYGGAAPGGPYGQPPPSSYGAQQPGLYGQGGAPPNVDPEAYSWFQSVDSDHSGYISMKELKQALVNCNWSSFNDETCLMMINMFDKTKSGRIDVYGFSALWKFIQQWKNLFQQYDRDRSGSISYTELQQALSQMGYNLSPQFTQLLVSRYCPRSANPAMQLDRFIQVCTQLQVLTEAFREKDTAVQGNIRLSFEDFVTMTASRML | |
| 50 μg | |
| Signal Transduction | |
| 553115 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| AAH02773 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction