missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEG10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554382
This item is not returnable.
View return policy
Description
Immunogen sequence: SIPNLEDQAH LATYTEFVPQ IPGYQTYPTY AAYPTYPVGF AWYPVGRDGQ GRSLYVPVMI TWNPHWYRQP PVPQYPPPQP PPPPPPPPPP PSYSTL Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 32%.
This is a paternally expressed imprinted gene that encodes transcripts containing two overlapping open reading frames start codon, have been reported for this gene. Increased expression of this gene is associated with hepatocellular carcinomas.
Specifications
| PEG10 | |
| Polyclonal | |
| Unconjugated | |
| PEG10 | |
| 2-Mar; AA407948; EDR; embryonal carcinoma differentiation regulated; embryonal carcinoma differentiation regulated protein; Embryonal carcinoma differentiation-regulated protein; HB-1; KIAA1051; Mammalian retrotransposon-derived protein 2; MAR2; Mart2; MEF3 like 1; MEF3L; Mef3l1; MEF3-like protein 1; MyEF-3; MyEF-3 like; myelin expression factor 3; Myelin expression factor 3-like protein 1; paternally expressed 10; paternally expressed gene 10 protein; PEG10; Retrotransposon gag domain-containing protein 3; retrotransposon-derived gag-like polyprotein; Retrotransposon-derived protein PEG10; retrotransposon-derived protein PEG10-like; RGAG3; Ty3/Gypsy-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23089 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86TG7 | |
| PEG10 | |
| Recombinant protein corresponding to Human PEG10. Recombinant protein control fragment (Product #RP-104311). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction