missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEG3 Antibody (OTI1E10), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73333AF647
This item is not returnable.
View return policy
Description
PEG3 Monoclonal specifically detects PEG3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PEG3 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PEG3 | |
| Recombinant protein fragment corresponding to amino acids 1-364 of human PEG3 (NP_006201) produced in E.coli. | |
| 0.1 mL | |
| 5178.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1E10 | |
| Western Blot, Immunohistochemistry | |
| DKFZp781A095, KIAA0287PW1, paternally expressed 3, paternally expressed gene 3, Zinc finger and SCAN domain-containing protein 24, ZNF904, ZSCAN24Kruppel-type zinc finger protein | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction