missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PELI1 Monoclonal Antibody (OTI13F8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807629S
This item is not returnable.
View return policy
Description
PELI1 Monoclonal antibody specifically detects PELI1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| PELI1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P18846 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 466 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI13F8 | |
| Unconjugated | |
| ATF1 | |
| 2810468L03Rik; A930031K15Rik; AA409794; AI586297; D11Ertd676e; E3 ubiquitin-protein ligase pellino homolog 1; Peli1; pellino 1; pellino E3 ubiquitin protein ligase 1; pellino homolog 1; pellino1; pellino-1; Pellino-related intracellular-signaling molecule; PRISM; protein pellino homolog 1; RING-type E3 ubiquitin transferase pellino homolog 1 | |
| Full length human recombinant protein of human ATF1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction