Learn More
PELI1 Monoclonal Antibody (OTI3A9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA807607S
Encompass_Preferred
 View more versions of this product
Product Code. 17598223
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
PELI1 Monoclonal antibody specifically detects PELI1 in Human samples. It is validated for Western Blot
Specifications
Specifications
PELI1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q96FA3
Mouse
Affinity chromatography
RUO
57162
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI3A9
Unconjugated
PELI1
2810468L03Rik; A930031K15Rik; AA409794; AI586297; D11Ertd676e; E3 ubiquitin-protein ligase pellino homolog 1; Peli1; pellino 1; pellino E3 ubiquitin protein ligase 1; pellino homolog 1; pellino1; pellino-1; Pellino-related intracellular-signaling molecule; PRISM; protein pellino homolog 1; RING-type E3 ubiquitin transferase pellino homolog 1
Human recombinant protein fragment corresponding to amino acids 1-245 of human PELI1 produced in E.coli.
30 μL