missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pellino 1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93523-0.02ml
This item is not returnable.
View return policy
Description
Pellino 1 Polyclonal antibody specifically detects Pellino 1 in Rat samples. It is validated for Western Blot
Specifications
| Pellino 1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DKFZp686C18116, MGC50990, pellino (Drosophila) homolog 1, pellino homolog 1 (Drosophila), pellino-1, Pellino-related intracellular-signaling molecule, PRISM, protein pellino homolog 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human PELI1 (NP_065702.2). MFSPDQENHPSKAPVKYGELIVLGYNGSLPNGDRGRRKSRFALFKRPKANGVKPSTVHIACTPQAAKAISNKDQHSISYTLSRAQTVVVEYTHDSNTDMFQIGRSTESPIDFVVTDTVPGSQSNSDTQSVQSTISRFACRIICERNPPFTARIYAAGFDSSKNIFLGEKAAKWKTSDGQMDGLTTNGVLVMHPRNGFTEDSKPGIWREISVCGNVFSLRETRSAQQRGKMVEIETNQLQDGSLIDLCGAT | |
| 0.02 mL | |
| Immunology, Signal Transduction | |
| 57162 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction