missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pellino 3 Monoclonal Antibody (K40052_17C1), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA555779

 View more versions of this product

Product Code. 30197680

  • € 438.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence of this protein is as follows: SPRTSDLQHR GNKGSCVLSS PGEDAQPGEE PIKYGELIVL GCCEEGGEET EAQRGEVTGP RAHSCYNGCL ASGDKGRRRS RLALSRRSHA NGVKPDVMHH ISTPLVSKAL SNRGQHSISY TLSRSHSVIV EYTHDSDTDM FQIGRSTENM IDFVVTDTSP GGGAAEGPSA

Pellino 1 is the mammalian homologue of Pellino, a Drosophila Pelle-interacting protein. Pelle is a serine-threonine kinase through which Toll signaling can occur. Pellino 1 can activate c-Jun, Elk-1 and NF-kB, whereas Pellino 3 can only activate c-Jun and Elk-1(2).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pellino 3
Monoclonal
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Q8N2H9
PELI3
Recombinant human PELI3 (immunogen range is 11-180).
50 μL
Primary
Human
Antibody
IgG1
Western Blot
K40052_17C1
Unconjugated
PELI3
6030441F14Rik; A930011L17; BC028931; E3 ubiquitin-protein ligase pellino homolog 3; E3 ubiquitin-protein ligase pellino homolog 3 pseudogene; MGC35521; PELI3; pellino 3; pellino E3 ubiquitin protein ligase family member 3; pellino homolog 3; pellino-3; protein pellino homolog 3; RGD1305989; RING-type E3 ubiquitin transferase pellino homolog 3
Mouse
Protein A
RUO
246330
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.