Learn More
PELO Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10582-1-AP-150UL
 View more versions of this product
Product Code. 16847053
  • € 410.00 / 150µL
Description
This gene encodes a protein which contains a conserved nuclear localization signal. The encoded protein may have a role in spermatogenesis, cell cycle control, and in meiotic cell division.
Specifications
Specifications
PELO
Polyclonal
Unconjugated
PELO
CGI 17, PELO, pelota homolog (Drosophila), PRO1770, Protein pelota homolog
Rabbit
Antigen Affinity Chromatography
RUO
105083, 294754, 53918
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5XIP1, Q80X73, Q9BRX2
PELO
PELO Fusion Protein Ag0873
150 μL
Primary
Human, Mouse, Rat