missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PELO Recombinant Rabbit Monoclonal Antibody (HL2345)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550497
This item is not returnable.
View return policy
Description
PELO Recombinant Monoclonal Antibody for Western Blot, IHC, IHC (P)
PELO encodes a protein which contains a conserved nuclear localization signal. The encoded protein may have a role in spermatogenesis, cell cycle control, and in meiotic cell division.
Specifications
| PELO | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q5XIP1, Q80X73, Q9BRX2 | |
| PELO | |
| Recombinant fragment of human PELO. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| HL2345 | |
| Unconjugated | |
| PELO | |
| AA409897; AI852218; CGI-17; PELO; pelota homolog (Drosophila); PRO1770; Protein pelota homolog | |
| Rabbit | |
| Protein A | |
| RUO | |
| 105083, 294754, 53918 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction