missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PELP1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-40622AF750
This item is not returnable.
View return policy
Description
PELP1 Polyclonal antibody specifically detects PELP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PELP1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Immunogen affinity purified | |
| RUO | |
| Store at 4C in the dark. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Rabbit | |
| 0.1 mL | |
| Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction