missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PELP1 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-40622B
This item is not returnable.
View return policy
Description
PELP1 Polyclonal antibody specifically detects PELP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PELP1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| HMX3, MNARmodulator of nongenomic activity of estrogen receptor, Modulator of non-genomic activity of estrogen receptor, P160, proline and glutamic acid rich nuclear protein, proline, glutamate and leucine rich protein 1, proline, glutamic acid and leucine rich protein 1, proline-, glutamic acid- and leucine-rich protein 1, proline-, glutamic acid-, leucine-rich protein 1, Transcription factor HMX3 | |
| The immunogen recognized by this antibody maps to a region between residue 25 and 75 of human Proline-, Glutamic Acid-, Leucine-Rich Protein 1 (Modulator of Nongenomic Activity of Estrogen Receptor) using the numbering given in entry AAN41255.1 (GeneID 27 | |
| 0.1 mL | |
| Cellular Markers, Transcription Factors and Regulators | |
| 27043 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction