Learn More
PELP1 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB200-331JF646
This item is not returnable.
View return policy
Description
PELP1 Polyclonal antibody specifically detects PELP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Immunohistochemistry | |
| Janelia Fluor 646 | |
| Primary |
| Polyclonal | |
| A synthetic peptide mapping to a region between residues 1000 and 1050 of human, proline-, glutamic acid-, leucine-rich protein 1/ modulator of nongenomic activity of estrogen receptor using the numbering given in entry AAN41255.1 (GeneID 27043). | |
| Purified |