Learn More
PELP1 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB200-331JF646
Product Code. 18695219
  • € 633.00 / 0.10mL
Description
PELP1 Polyclonal antibody specifically detects PELP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunohistochemistry
Janelia Fluor 646
Primary
Polyclonal
A synthetic peptide mapping to a region between residues 1000 and 1050 of human, proline-, glutamic acid-, leucine-rich protein 1/ modulator of nongenomic activity of estrogen receptor using the numbering given in entry AAN41255.1 (GeneID 27043).