Learn More
3M™ PELTOR™ Bull's Eye™ I Earmuffs
Catalog Number 15739707
Click to view available options
Color:
Black
Green
Red
3M™ PELTOR™ Bull's Eye™ I Earmuffs ensure protection in high noise environments without being too bulky.
  • Shaped to avoid interference with the rifle butt and not distract when shooting
  • Soft wide cushions helps reduce pressure around the ears and improves comfort and wearability
  • Easy to replace cushions and inserts helps keep them hygienically clean
  • Collapsible headband for easy and protective storage in a pocket or bag
  • Attenuation Rating:
    • High: 32
    • Medium: 24
    • Low: 15
Color Red
Certifications/Compliance CE Approved , EN 352-1: 2002
Noise Reduction Rating (NRR) 27 dB
Product Type Ear Muffs
Style Foldable, over the head
Foldable Yes