Learn More
3M™ PELTOR™ Optime™ II Earmuffs
Catalog Number 15739697
Click to view available options
Model:
H520A-472-GB
H520F-409-GQ
H520F-460-GB
H520P3A-410-GQ
H520P3AF-410-GQ
H520P3B-410-GQ
H520P3E-410-GQ-01
H520P3E-467-GB
H520P3EA-410-GQ
H520P3F-410-GQ
H520P3G-410-GQ
H520P3H-410-GQ
H520P3H-467-GB
H520P3K-410-GQ
Ideal for environments with substantial industrial noise or construction machinery, like airports and agricultural work. 3M™ PELTOR™ Optime™ II Earmuffs have been developed for demanding noise-hazard environments.
  • Ideal hearing protection against noise arising from environments with substantial industrial noise or construction machinery, like airports and agricultural work
  • Soft wide cushions helps reduce pressure around the ears and improves comfort and wearability
  • Large space inside cup helps reduce moisture and heat build-up
  • Easy to replace cushions and inserts helps keep them hygienically clean
Color Hi-Viz
Certifications/Compliance CE Approved, EN 352-3: 2002
Model H520F-460-GB
Noise Reduction Rating (NRR) 31 dB
Product Type Ear Muffs
Dielectric No
Foldable Yes