missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pemetrexed disodium, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
Description
Pemetrexed disodium (LY231514 disodium) is an antifolate, the Kis of the pentaglutamate of Pemetrexed disodium are 1.3, 7.2, and 65 nM for inhibits thymidylate synthase (TS), dihydrofolate reductase (DHFR), and glycinamide ribonucleotide formyltransferase (GARFT), respectively.
Specifications
Specifications
| Chemical Name or Material | Pemetrexed disodium |
| CAS | 150399-23-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H19N5Na2O6 |
| Quantity | 100 mg |
| Synonym | LY231514 disodium |
| Solubility Information | H2O : ≥ 100 mg/mL (212.15 mM) |
| SMILES | O=C1NC(N)=NC2=C1C(CCC3=CC=C(C=C3)C(N[C@@H](CCC(O[Na])=O)C(O[Na])=O)=O)=CN2 |
| Molecular Weight (g/mol) | 471.37 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction