missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PEN2 Polyclonal Antibody

Product Code. 11578801
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11578801 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11578801 Supplier Invitrogen™ Supplier No. PA316762

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody has primarily been tested in HA-PEN2 transfected cells combining Immunoprecipitation with Western blot analysis (ECL). For endogenous sample testing, it is suggested that the investigator load a greater amount of protein on the gel. Suggested positive control: antigen standard for PSENEN (transient overexpression lysate).

Presenilins, which are components of the gamma-secretase protein complex, are required for intramembranous processing of some type I transmembrane proteins, such as the Notch proteins and the beta-amyloid precursor protein. Signaling by Notch receptors mediates a wide range of developmental cell fates. Processing of the beta-amyloid precursor protein generates neurotoxic amyloid beta peptides, the major component of senile plaques associated with Alzheimer's disease. This gene encodes a protein that is required for Notch pathway signaling, and for the activity and accumulation of gamma-secretase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEN2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.05% sodium azide
Gene PSENEN
Gene Accession No. Q9CQR7, Q9NZ42
Gene Alias 1700023M09Rik; gamma-secretase subunit PEN-2; hematopoietic stem/progenitor cells protein MDS033; liver regeneration-related protein LRRGT00140; LRRGT00140 mRNA; MDS033; MSTP064; Pen2; Pen-2; presenilin enhancer 2; presenilin enhancer 2 homolog; presenilin enhancer 2 homolog (C. elegans); presenilin enhancer gamma secretase subunit; presenilin enhancer gamma-secretase subunit; presenilin enhancer protein 2; presenilin enhancer, gamma-secretase subunit; Psenen; RGD1312037
Gene Symbols PSENEN
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus of human PEN2 (within residues 50-101).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55851, 66340
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.