missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Penconazole Monoclonal Antibody (3G10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549661
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Antibody tested in indirect ELISA.
Penconazole is a fungicide that belongs to the chemical class of triazoles. It is used to control fungal diseases in crops such as cereals, vegetables, fruits, and ornamentals. Penconazole works by inhibiting the growth of fungi by interfering with the production of ergosterol, an essential component of the fungal cell membrane. It is often used in combination with other fungicides to increase its effectiveness and prevent the development of resistant strains of fungi.
Spezifikation
| Penconazole | |
| Monoclonal | |
| 2.22 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Conjugate of PCZ hapten and KLH. | |
| 200 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry, Lateral Flow, SDS-Page | |
| 3G10 | |
| Unconjugated | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur