Learn More
Pendimethalin-d5 [N-(1-ethyl-1',1'-d2-propyl-1,2,2-d3)], CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Pendimethalin D5 |
| CAS | 1219803-39-0 |
| Molecular Formula | C13 2H5 H14 N3 O4 |
| Quantity | 0.01 g |
| Synonym | 3,4-Dimethyl-2,6-dinitro-N-(2,2,3,4,4-pentadeuteriopentan-3-yl)aniline, Pendimethalin D5 (1-Ethyl(1',1'-D2)propyl(1,2,2-D3)), Pendimethalin D5 (pent-3-yl (2,2,3,4,4)-D5) |
| InChI Formula | InChI=1 S/C13H19N3O4/c1-5-10(6-2)14-12-11(15(17)18)7-8(3)9(4)13(12)16(19)20/h7,10,14 H,5-6H2,1-4H3/i5D2,6D2,10 D |
| SMILES | [2 H]C([2 H])(C)C([2 H])(Nc1c(cc(C)c(C)c1[N+](=O)[O-])[N+](=O)[O-])C([2 H])([2 H])C |
| IUPAC Name | 3,4-dimethyl-2,6-dinitro-N-(2,2,3,4,4-pentadeuteriopentan-3-yl)aniline |
| Molecular Weight (g/mol) | 286.34 |
| Formula Weight | 286.1689 g/mol |
| Show More |