missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Penitrem A

Description
Potent and selective blocker of BKCa (KCa1.1) channels (IC50 values are 6.4 and 64.4 nM for BKCa channels containing α subunits only, and those containing α and β1 respectively). Displays no effect on native delayed rectifier K+ and KATP currents, or cloned KV1.5 channels. Blocks BKCa channels in both inside-out and cell-attached patches. Shown to enhance smooth muscle contraction in vitro and increase total peripheral resistance in vivo.
Specifications
Specifications
| CAS | 12627-35-9 |
| Quantity | 1 mg |
| Molecular Formula | C37H44ClNO6 |
| Formula Weight | 634.2 |
| Purity | >98% |
| Molecular Weight (g/mol) | 634.21 |
| Chemical Name or Material | Penitrem A |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?