missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ PENK Polyclonal Antibody

Artikelnummer. 16508291
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
100 μL
Pakningsstørrelse:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
16508291 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16508291 Leverandør Invitrogen™ Leverandørnr. PA592882

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

Positive Samples: Rat spinal cord, Rat brain.

neuropeptide hormone; plays a role in the responses to stress and pain perception.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen PENK
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene PENK
Gene Accession No. P01210, P04094, P22005
Gene Alias AI326464; ENK; enkephalin; enkephalin A; Leu-enkephalin; Met-enkephalin; Met-enkephalin-Arg-Gly-Leu; Met-enkephalin-Arg-Phe; Met-enkephalin-Arg-Ser-Leu; OGF; Opioid growth factor; PE; Penk; PENK(114-133); PENK(143-183); PENK(143-184); PENK(143-185); PENK(237-258); PENK(238-259); PENK(239-260); Penk1; Penk2; PENK-A; Penk-rs; PPA; preproenkephalin; preproenkephalin 1; preproenkephalin 2; proenkephalin; proenkephalin 1; proenkephalin 2; proenkephalin related sequence; proenkephalin-A; spermatogenic-specific proenkephalin; Synenkephalin
Gene Symbols PENK
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18619, 29237, 5179
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.