missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluoropyridine, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Pentafluoropyridine was used in the preparation of η2-C,C coordinated pentafluoropyridine complex. It was also used in the preparation of polyfluorinated coupling products via Pd(0)-catalyzed cross-coupling reaction with diarylzinc compounds. It was also used as derivatizing reagent in the sensitive GC-MS method for determination of the four endocrine disrupting chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pentafluoropyridine was used in the preparation of η2-C,C coordinated pentafluoropyridine complex. It was also used in the preparation of polyfluorinated coupling products via Pd(0)-catalyzed cross-coupling reaction with diarylzinc compounds. It was also used as derivatizing reagent in the sensitive GC-MS method for determination of the four endocrine disrupting chemicals.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents, alkali metals and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Pentafluoropyridine was used in the preparation of η2-C,C coordinated pentafluoropyridine complex. It was also used in the preparation of polyfluorinated coupling products via Pd(0)-catalyzed cross-coupling reaction with diarylzinc compounds. It was also used as derivatizing reagent in the sensitive GC-MS method for determination of the four endocrine disrupting chemicals.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents, alkali metals and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Density | 1.618 |
Boiling Point | 83°C to 85°C |
Flash Point | 23°C (73°F) |
Refractive Index | 1.3865 |
Quantity | 5 g |
UN Number | UN2924 |
Beilstein | 1427064 |
Solubility Information | Not miscible or difficult to mix in water. |
Formula Weight | 169.05 |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'