Learn More
Pentanoic-2,2,3,3-d4 Acid, TRC
Product Code. 30457297
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457297 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30457297 Supplier Toronto Research Chemicals Supplier No. TRCV91417110MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Pentanoic-2,2,3,3-d4 Acid
CAS 1219804-71-3
Molecular Formula C5 D4 H6 O2
Quantity 10 mg
Synonym Pentanoic-2,2,3,3-d4 acid, Pentanoic acid-2,2,3,3-d4, Valeric acid-d4, Valeric-α,α,β,β-d4 acid, Pentanoic-2,2,3,3-D4 acid
InChI Formula InChI=1S/C5H10O2/c1-2-3-4-5(6)7/h2-4H2,1H3,(H,6,7)/i3D2,4D2
SMILES [2H]C([2H])(CC)C([2H])([2H])C(=O)O
IUPAC Name 2,2,3,3-tetradeuteriopentanoic acid
Molecular Weight (g/mol) 106.156
Formula Weight 106.093