missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentanoic-d9 Acid, CDN
Specifications
Specifications
| Chemical Name or Material | Valeric Acid-d9 |
| CAS | 115871-50-6 |
| Molecular Formula | C5 D9 H O2 |
| Quantity | 5 g |
| Synonym | 1-Butanecarboxylic Acid-d9, NSC 406833-d9, Propylacetic Acid-d9, n-Pentanoic Acid-d9, n-Valeric Acid-d9, Pentanoic-d9 acid (9 CI), Pentanoic-d9 acid (9 CI), Pentanoic-d9 acid (9 CI), Pentanoic-d9 acid (9 CI) |
| InChI Formula | InChI=1 S/C5H10O2/c1-2-3-4-5(6)7/h2-4H2,1H3,(H,6,7)/i1D3,2D2,3D2,4D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)O |
| IUPAC Name | 2,2,3,3,4,4,5,5,5-nonadeuteriopentanoic acid |
| Molecular Weight (g/mol) | 111.19 |
| Formula Weight | 111.1246 g/mol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?