missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentraxin 3/TSG-14 Antibody (10D9), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | Pentraxin 3/TSG-14 |
|---|---|
| Clone | 10D9 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30039714
|
Novus Biologicals
NBP3-26356-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30038313
|
Novus Biologicals
NBP3-26356-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pentraxin 3/TSG-14 Monoclonal antibody specifically detects Pentraxin 3/TSG-14 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| Pentraxin 3/TSG-14 | |
| Flow Cytometry 1:50-1:200, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| alpha-induced protein 5, pentaxin-related gene, rapidly induced by IL-1 beta, tumor necrosis factor, Pentaxin-related protein PTX3, pentraxin 3, long, pentraxin-3, pentraxin-related gene, rapidly induced by IL-1 beta, pentraxin-related protein PTX3, TNF alpha-induced protein 5, TNFAIP5, TSG14, TSG-14pentaxin-related gene, rapidly induced by IL-1 beta, Tumor necrosis factor alpha-induced protein 5, tumor necrosis factor, alpha-induced protein 5, Tumor necrosis factor-inducible gene 14 protein, tumor necrosis factor-inducible protein TSG-14 | |
| A synthesized peptide derived from Human Pentraxin 3/TSG-14 [UniProt P26022] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 10D9 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Immunology | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 5806 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title