missing translation for 'onlineSavingsMsg'
Learn More

PEPA, MedChemExpress

Product Code. 16481437
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481437 50 mg 50mg
16445228 10 mg 10mg
16437947 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16481437 Supplier Medchem Express Supplier No. HY12509

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PEPA is an allosteric modulator of AMPA receptors; binds to the GluA2o and GluA3o LBDs and can be utilized as an indicator of AMPA receptor heterogeneity.

PEPA is an allosteric modulator of AMPA receptors

Specifications

Chemical Name or Material PEPA
CAS 141286-78-4
Color White
Purity Grade Notes Research
Molecular Formula C16H16F2N2O4S2
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (124.24 mM; Need ultrasonic)
SMILES FC1=CC(SCCNS(C2=CC=CC=C2)(=O)=O)=CC(F)=C1OCC(N)=O
Molecular Weight (g/mol) 402.44
Formula Weight 402.44
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.