Learn More
Pepsin Antibody, HRP, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB120-20732-0.1mg
This item is not returnable.
View return policy
Description
Pepsin Polyclonal antibody specifically detects Pepsin in Porcine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Pepsin | |
| Polyclonal | |
| HRP | |
| Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free | |
| DKFZp666J2410, ec 3.4.23.1, Pepsin a precursor, Pepsinogen 1 second locus, Pepsinogen 3 group 1, Pepsinogen 4 group I, Pepsinogen 5 group I, Pepsinogen A, Pepsinogen a3, Pepsinogen A4, Pepsinogen A5, Pepsinogen group 1, Pepsinogen I, Pga 3, Pga 4, Pga 5, pga3, pga4, pga5 | |
| Pepsin Antibody was produced by repeated immunizations with pork Pepsin protein isolated from swine stomach. (Uniprot: P00791) | |
| 0.1 mg | |
| Primary | |
| Pig | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot 1:500-1:2500, ELISA 1:2000-1:10000, Immunohistochemistry 1:250-1:1250, Immunohistochemistry-Paraffin 1:10-1:500 | |
| P00791 | |
| Goat | |
| Multi-step | |
| RUO | |
| Reconstitute with 100 μg deionized water (or equivalent) | |
| Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. |