Learn More
Pepsin Antibody, HRP, Novus Biologicals™
Goat Polyclonal Antibody
Brand:  Novus Biologicals NB120-20732-0.1mg
Product Code. 18544973
  • € 644.00 / 0.10mg
Description
Pepsin Polyclonal antibody specifically detects Pepsin in Porcine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pepsin
Polyclonal
HRP
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free
DKFZp666J2410, ec 3.4.23.1, Pepsin a precursor, Pepsinogen 1 second locus, Pepsinogen 3 group 1, Pepsinogen 4 group I, Pepsinogen 5 group I, Pepsinogen A, Pepsinogen a3, Pepsinogen A4, Pepsinogen A5, Pepsinogen group 1, Pepsinogen I, Pga 3, Pga 4, Pga 5, pga3, pga4, pga5
Pepsin Antibody was produced by repeated immunizations with pork Pepsin protein isolated from swine stomach. (Uniprot: P00791)
0.1 mg
Primary
Pig
IgG
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
LYOPH
Western Blot 1:500-1:2500, ELISA 1:2000-1:10000, Immunohistochemistry 1:250-1:1250, Immunohistochemistry-Paraffin 1:10-1:500
P00791
Goat
Multi-step
RUO
Reconstitute with 100 μg deionized water (or equivalent)
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.