missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pepsinogen A Antibody (3D1C6), HRP , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06077H
This item is not returnable.
View return policy
Description
Pepsinogen A Monoclonal antibody specifically detects Pepsinogen A in Human samples. It is validated for ELISA
Specifications
| Pepsinogen A | |
| Monoclonal | |
| HRP | |
| EC 3.4.23, EC 3.4.23.1, FLJ58952, FLJ77962, pepsin A, pepsinogen 4, group I (pepsinogen A), pepsinogen A4, PGA3, PGA5 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Pepsinogen A (Met 1-Ala 388). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 3D1C6 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 643847 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction