missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pepsinogen I Antibody (8003 (99/12)), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62239AF647
This item is not returnable.
View return policy
Description
Pepsinogen I Monoclonal specifically detects Pepsinogen I in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Pepsinogen I | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PGA3 | |
| Purified human pepsinogen 1. | |
| 0.1 mL | |
| Primary | |
| NB100-62239 recognises human Pepsinogen I, a zymogen or proenzyme secreted by Chief cells in the stomach. It is cleaved to form pepsin both in an autocatalytic fashion and by pepsin itself. In humans there are two related forms of pepsin, Pepsinogen I (also known as pepsinogen A), and Pepsinogen II (also known as Pepsinogen B or progastricsin). | |
| Store at 4C in the dark. | |
| IgG1 |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 8003 (99/12) | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| pepsinogen 3, group I (pepsinogen A) | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 643834 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction