missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEPT1/SLC15A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98544-100ul
This item is not returnable.
View return policy
Description
PEPT1/SLC15A1 Polyclonal antibody specifically detects PEPT1/SLC15A1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| PEPT1/SLC15A1 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Caco-2 oligopeptide transporter, HPECT1macrophage oligopeptide transporter PEPT1, HPEPT1, Intestinal H(+)/peptide cotransporter, Oligopeptide transporter, small intestine isoform, PEPT1intestinal H+/peptide cotransporter, Peptide transporter 1, solute carrier family 15 (oligopeptide transporter), member 1, solute carrier family 15 member 1 | |
| Produced in rabbits immunized with E. coli-derived Human PEPT1/SLC15A1 fragment. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 6564 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction