missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peptide YY Antibody (1), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05167JF549
This item is not returnable.
View return policy
Description
Peptide YY Monoclonal specifically detects Peptide YY in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PeptideYY | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PYY | |
| Synthetic human PYY adsorbed onto aluminum hydroxide gel | |
| 0.1 mL | |
| Primary | |
| Binds human PYY. Cross-reactivity with NPY or PP in solution is | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Peptide tyrosine tyrosine, peptide YY, PYY1, PYY-I | |
| Mouse | |
| Protein A or G purified | |
| Signal Transduction | |
| 5697 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction