missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PER1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 271.00 - € 568.00
Specifications
| Antigen | PER1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18281961
|
Novus Biologicals
NBP2-57816 |
100 μL |
€ 568.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18644417
|
Novus Biologicals
NBP2-57816-25ul |
25 μL |
€ 271.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PER1 Polyclonal specifically detects PER1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| PER1 | |
| Polyclonal | |
| Rabbit | |
| Circadian Rhythm, Neuroscience, Vision | |
| Circadian clock protein PERIOD 1, Circadian pacemaker protein Rigui, hPER1, KIAA0482, PERhPER, period (Drosophila) homolog 1, period circadian protein homolog 1, period homolog 1 (Drosophila), Period, drosophila, homolog of, RIGUIMGC88021 | |
| PER1 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 5187 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LEGADGGGDPRPGESFCPGGVPSPGPPQHRPCPGPSLADDTDANSNGSSGNESNGHESRGASQRSSHSSSSGNGKDSALLETTE | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title