missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PER2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94634-0.02ml
This item is not returnable.
View return policy
Description
PER2 Polyclonal antibody specifically detects PER2 in Human, Rat samples. It is validated for Western Blot
Specifications
| PER2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| Circadian clock protein PERIOD 2, FASPS, hPER2, KIAA0347period 2, period (Drosophila) homolog 2, period circadian protein 2, period circadian protein homolog 2, period homolog 2 (Drosophila) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 390-565 of human PER2 (NP_073728.1). GQPFDYSPIRFRARNGEYITLDTSWSSFINPWSRKISFIIGRHKVRVGPLNEDVFAAHPCTEEKALHPSIQELTEQIHRLLLQPVPHSGSSGYGSLGSNGSHEHLMSQTSSSDSNGHEDSRRRRAEICKNGNKTKNRSHYSHESGEQKKKSVTEMQTNPPAEKKAVPAMEKDSLGV | |
| 0.02 mL | |
| Cancer, Chromatin Research, Circadian Rhythm, Endothelial Cell Markers, Hypoxia, Lipid and Metabolism, Neuroscience, Signal Transduction, Transcription Factors and Regulators, Vision | |
| 8864 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction