Learn More
PER2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29954-1-AP-150UL
This item is not returnable.
View return policy
Description
The PER2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PER2. This antibody recognizes human antigen. The PER2 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| PER2 | |
| 750 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, Immunofluorescence, ELISA | |
| FASPS, hPER2, KIAA0347, PER2, period homolog 2 (Drosophila) | |
| 150 μL | |
| 8864 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |