Learn More
Peramivir trihydrate, MedChemExpress
Product Code. 16457268
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 16457268
Brand: Medchem Express HY17015
MedChemExpress Peramivir trihydrate (RWJ-270201 trihydrate;BCX-1812 trihydrate) is a highly potent, selective and orally active influenza virus neuraminidase (NA) inhibitor, with IC50 values ranging from 0.9 to 4.3 nM for nine NA subtypes.
Peramivir trihydrate (RWJ-270201 trihydrate
Specifications
Chemical Name or Material Peramivir trihydrate
Physical Form Powder
CAS 1041434-82-5
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C15H34N4O7
For Use With (Application) COVID-19-anti-virus
Synonym RWJ 270201 trihydrate BCX 1812 trihydrate
Solubility Information H2O : 5 mg/mL (13.07 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (ultrasonic) (insoluble or slightly soluble)
SMILES O[C@H]1[C@]([C@H](C(CC)CC)NC(C)=O)([H])[C@H](NC(N)=N)C[C@@H]1C(O)=O.[H]O.[H]O.[H]O
Molecular Weight (g/mol) 382.45
Formula Weight 382.45
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only