missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid, 0.1M, Chem Lab™

Brand:  Chem Lab CL05.1602.1000

Additional Details : CAS : 64-19-7 Weight : 1.25000kg Transport : UN number : 2789 Chem class : 8 Pack group : II


Quantity 1L

 View more versions of this product

Product Code. 11973263

  • € 80.15 / 1 L


Estimated Shipment: 01-09-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

Specifications

Colorless
<2
Liquid
1L
1L
1.05g/mL
Acetic Acid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Perchloric acid 0.1 mol/l in Acetic acid 100%

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : perchloric acid, acetic acid

SDS
Documents
Certificates