Learn More
Perfluoro-compound FC-770TM
Product Code. 10412785 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
50 mL
Unit Size:
25mL
50mL
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10412785
Brand: Thermo Scientific Acros 394890250
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
CAS: 1093615-61-2399 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1093615-61-2
Molecular Weight (g/mol) 399
MDL Number MFCD09753882
Synonym Fluorinert™ FC-770
Specifications
Density 1.8000g/mL
Boiling Point 95.0°C
Packaging Glass bottle
Quantity 25 mL
Specific Gravity 1.8
Solubility Information Solubility in water: insoluble
Viscosity 0.8 mm2/s (25°C)
Formula Weight 399
Chemical Name or Material Perfluoro-compound FC-770*
Product Identifier
  • Perfluoro-compound FC-770]TS
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
Hazard Statement
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.