Learn More
Perfluoro-compound FC-770TM
Product Code. 10704184 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
50 mL
Unit Size:
25mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10704184 50 mL 50mL
10412785 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10704184 Supplier Thermo Scientific Acros Supplier No. 394890500
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
CAS: 1093615-61-2399 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1093615-61-2
Molecular Weight (g/mol) 399
MDL Number MFCD09753882
Synonym Fluorinert™ FC-770
Specifications
Density 1.8000g/mL
Boiling Point 95.0°C
Packaging Glass bottle
Quantity 50 mL
Specific Gravity 1.8
Solubility Information Solubility in water: insoluble
Viscosity 0.8 mm2/s (25°C)
Formula Weight 399
Chemical Name or Material Perfluoro-compound FC-770*
Product Identifier
  • Perfluoro-compound FC-770]TS
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
Hazard Statement
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.